Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Heyl
Entrez Gene ID
56198
Gene Name
hairy/enhancer-of-split related with YRPW motif-like
Gene Aliases
Hey3, Hrt3, bHLHb33, hesr3
Chromosome Location
Chr.4: 123233556 - 123249870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013905.3 NP_038933.2 1 - 2 394 83
GenBank mRNA AB093590.1 - 1 - 2 86 83
AF172288.1 - 1 - 2 394 83
AJ271868.2 - 1 - 2 394 83
AK028596.1 - 1 - 2 133 83
AK080822.1 - 1 - 2 98 83
AK165078.1 - 1 - 2 100 83
BC130263.1 - 1 - 2 102 83

View Full Product Details