The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015769.2 | NP_056584.2 | 1 - 2 | 407 | 87 |
GenBank mRNA | AF189285.1 | - | 1 - 2 | 407 | 87 |
AK018375.1 | - | 1 - 2 | 227 | 87 | |
AK043366.1 | - | 1 - 2 | 209 | 87 | |
AK138588.1 | - | 1 - 2 | 201 | 87 | |
AK149326.1 | - | 1 - 2 | 240 | 87 | |
BC026792.1 | - | 1 - 2 | 198 | 87 |