The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016669.1 | NP_057878.1 | 7 - 8 | 941 | 70 |
GenBank mRNA | AF039391.1 | - | 7 - 8 | 941 | 70 |
AK138422.1 | - | 7 - 8 | 912 | 70 | |
AK143116.1 | - | 7 - 8 | 931 | 70 | |
AK163781.1 | - | 7 - 8 | 912 | 70 | |
BC045159.1 | - | 7 - 8 | 902 | 70 |