Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coil
Entrez Gene ID
12812
Gene Name
coilin
Gene Aliases
C79982, Cln80, p80, p80-coilin
Chromosome Location
Chr.11: 88973935 - 88991613 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016706.2 NP_057915.2 3 - 4 1459 92
GenBank mRNA AF208663.1 - 3 - 4 1441 92
AK146366.1 - 3 - 4 1443 92
BC031167.1 - 3 - 4 1474 92
CF105200.1 - 2 - 3 257 92

View Full Product Details