The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AB009384.1 | - | 2 - 3 | 75 | |
AF030185.1 | - | 1 - 2 | 75 | ||
AF316576.1 | - | 1 - 2 | 206 | 75 | |
AJ318863.1 | - | 1 - 2 | 75 | ||
AK007808.1 | - | 1 - 2 | 504 | 75 | |
AK145946.1 | - | 1 - 2 | 479 | 75 | |
AK151856.1 | - | 2 - 3 | 255 | 75 | |
BC038631.1 | - | 2 - 3 | 75 | ||
BY162594.1 | - | 2 - 3 | 253 | 75 |