The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017468.3 | NP_059496.1 | 8 - 9 | 847 | 63 |
GenBank mRNA | BC115848.1 | - | 8 - 9 | 759 | 63 |
U82698.1 | - | 8 - 9 | 848 | 63 |