Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Calcrl
Entrez Gene ID
54598
Gene Name
calcitonin receptor-like
Gene Aliases
AV071593, CRLR
Chromosome Location
Chr.2: 84330626 - 84425411 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018782.2 NP_061252.2 13 - 14 1423 105
GenBank mRNA AB015595.1 - 13 - 14 1427 105
AF146525.1 - 11 - 12 1100 105
AF209905.1 - 9 - 10 919 105
AK137210.1 - 13 - 14 1422 105
BC138537.1 - 10 - 11 1050 105
BC138540.1 - 10 - 11 1050 105

View Full Product Details