The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018782.2 | NP_061252.2 | 15 - 16 | 1677 | 121 |
GenBank mRNA | AB015595.1 | - | 15 - 16 | 1681 | 121 |
AF146525.1 | - | 13 - 14 | 1354 | 121 | |
AF209905.1 | - | 11 - 12 | 1173 | 121 | |
AK137210.1 | - | 15 - 16 | 1676 | 121 | |
BC138537.1 | - | 12 - 13 | 1304 | 121 | |
BC138540.1 | - | 12 - 13 | 1304 | 121 |