Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chm
Entrez Gene ID
12662
Gene Name
choroidermia (RAB escort protein 1)
Gene Aliases
Rep-1
Chromosome Location
Chr.X: 113040590 - 113185539 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018818.2 NP_061288.2 8 - 9 1222 68
GenBank mRNA AF218084.1 - 8 - 9 1231 68
AK035956.1 - 8 - 9 1217 68
AK141830.1 - 7 - 8 1100 68
BC047461.1 - 8 - 9 1221 68

View Full Product Details