Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2b2
Entrez Gene ID
23921
Gene Name
SH2B adaptor protein 2
Gene Aliases
Aps
Chromosome Location
Chr.5: 136218147 - 136246612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF234838.1 - 2 - 3 990 89
AK017604.1 - 2 - 3 960 89
AK140319.1 - 2 - 3 956 89
AK140325.1 - 2 - 3 956 89
BC049777.1 - 2 - 3 941 89
BC057334.1 - 2 - 3 1008 89

View Full Product Details