The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019517.4 | NP_062390.3 | 8 - 9 | 1364 | 64 |
GenBank mRNA | AF051150.1 | - | 4 - 5 | 513 | 64 |
AF216310.1 | - | 8 - 9 | 1315 | 64 | |
AK052309.1 | - | 8 - 9 | 1351 | 64 | |
AK078770.1 | - | 8 - 9 | 1331 | 64 | |
AK165036.1 | - | 8 - 9 | 1300 | 64 | |
BC120773.1 | - | 8 - 9 | 1330 | 64 | |
BC131947.1 | - | 8 - 9 | 1330 | 64 |