Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rapgef4
Entrez Gene ID
56508
Gene Name
Rap guanine nucleotide exchange factor (GEF) 4
Gene Aliases
1300003D15Rik, 5730402K07Rik, 6330581N18Rik, EPAC 2, Epac-2, Epac2, mKIAA4040
Chromosome Location
Chr.2: 71981215 - 72257474 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204165.1 NP_001191094.1 26 - 27 2726 67
NM_001204166.1 NP_001191095.1 23 - 24 2618 67
NM_019688.2 NP_062662.1 25 - 26 2672 67
GenBank mRNA AB021132.1 - 26 - 27 2701 67
AB037668.1 - 17 - 18 2061 67
AB445011.1 - 23 - 24 2610 67
AF115480.1 - 25 - 26 2600 67
AK004874.1 - 17 - 18 1718 67
AK220281.1 - 26 - 27 2726 67
BC139822.1 - 26 - 27 2658 67

View Full Product Details