Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fzr1
Entrez Gene ID
56371
Gene Name
fizzy/cell division cycle 20 related 1 (Drosophila)
Gene Aliases
AW108046, Cdh1, FZR, FZR2, Fyr, HCDH, HCDH1
Chromosome Location
Chr.10: 81366879 - 81378370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019757.1 NP_062731.1 2 - 3 208 66
GenBank mRNA AF083809.1 - 2 - 3 208 66
AK051993.1 - 2 - 3 290 66
AK081476.1 - 2 - 3 261 66
AK083686.1 - - 271 66
AK154822.1 - 2 - 3 292 66
AK155370.1 - 2 - 3 290 66
AK163951.1 - 2 - 3 267 66
AK172679.1 - 2 - 3 302 66
BC006616.1 - 2 - 3 289 66

View Full Product Details