The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190852.1 | NP_001177781.1 | 14 - 15 | 107 | |
NM_001190854.1 | NP_001177783.1 | 11 - 12 | 107 | ||
NM_001190855.1 | NP_001177784.1 | 12 - 13 | 107 | ||
NM_001190856.1 | NP_001177785.1 | 13 - 14 | 107 | ||
NM_019808.3 | NP_062782.2 | 11 - 12 | 107 | ||
GenBank mRNA | AB016586.1 | - | 11 - 12 | 107 | |
AK089779.1 | - | 11 - 12 | 1729 | 107 | |
AK147999.1 | - | 11 - 12 | 107 | ||
BC020145.1 | - | 3 - 4 | 348 | 107 | |
BC037476.1 | - | 11 - 12 | 107 | ||
DQ177283.1 | - | 10 - 11 | 1566 | 107 | |
HM369794.1 | - | 10 - 11 | 107 | ||
HM369795.1 | - | 11 - 12 | 107 | ||
HM369796.1 | - | 12 - 13 | 107 | ||
HM369797.1 | - | 13 - 14 | 107 |