The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020567.2 | NP_065592.1 | 6 - 7 | 591 | 84 |
GenBank mRNA | AF068780.1 | - | 6 - 7 | 738 | 84 |
AK131959.1 | - | 6 - 7 | 565 | 84 | |
BC020061.1 | - | 6 - 7 | 743 | 84 | |
CF751770.1 | - | 1 - 2 | 76 | 84 |