Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jph3
Entrez Gene ID
57340
Gene Name
junctophilin 3
Gene Aliases
JP-3, Jp3
Chromosome Location
Chr.8: 121730559 - 121791083 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020605.3 NP_065630.1 4 - 5 2219 65
GenBank mRNA AB024449.1 - 4 - 5 2215 65
AK045706.1 - 4 - 5 2206 65
AK046136.1 - - 2212 65
AK082880.1 - 4 - 5 2208 65
BC103682.1 - 4 - 5 2174 65
BC104736.1 - 4 - 5 2174 65
BC105307.1 - 4 - 5 2174 65

View Full Product Details