Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec5a
Entrez Gene ID
23845
Gene Name
C-type lectin domain family 5, member a
Gene Aliases
Clecsf5, Ly100, MDL-1, Mdl1
Chromosome Location
Chr.6: 40574898 - 40585805 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021364.2 NP_067339.1 2 - 3 263 129
GenBank mRNA AF139769.1 - 2 - 3 233 129
AK036697.1 - 2 - 3 229 129
AK046600.1 - 2 - 3 250 129
AK137482.1 - 2 - 3 264 129
BY205833.1 - 2 - 3 279 129

View Full Product Details