The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021487.1 | NP_067462.1 | 1 - 1 | 1049 | 86 |
GenBank mRNA | AJ131398.1 | - | 1 - 1 | 1049 | 86 |
AK005246.1 | - | 1 - 1 | 86 | ||
AK014537.1 | - | 1 - 1 | 883 | 86 | |
BC028942.1 | - | 1 - 1 | 1027 | 86 |