Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jph2
Entrez Gene ID
59091
Gene Name
junctophilin 2
Gene Aliases
1110002E14Rik, JP-2, Jp2
Chromosome Location
Chr.2: 163336242 - 163398049 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205076.1 NP_001192005.1 5 - 6 2374 82
NM_021566.2 NP_067541.1 4 - 5 2454 82
GenBank mRNA AB024447.1 - 4 - 5 2431 82
AK157145.1 - 5 - 6 2374 82
AY974567.1 - 1 - 2 196 82

View Full Product Details