Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fign
Entrez Gene ID
60344
Gene Name
fidgetin
Gene Aliases
fi, fidget
Chromosome Location
Chr.2: 63971508 - 64098038 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021716.5 NP_068362.1 2 - 3 365 109
GenBank mRNA AF263913.1 - 2 - 3 315 109
AK143787.1 - 2 - 3 340 109
AK163474.1 - 2 - 3 336 109

View Full Product Details