Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fads3
Entrez Gene ID
60527
Gene Name
fatty acid desaturase 3
Gene Aliases
AI464531
Chromosome Location
Chr.19: 10040855 - 10059671 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021890.3 NP_068690.3 3 - 4 720 81
GenBank mRNA AB041560.1 - 3 - 4 662 81
AK029318.1 - 3 - 4 579 81
AK080414.1 - 3 - 4 572 81
AK090042.1 - 3 - 4 702 81
AK154534.1 - 3 - 4 597 81
AK172509.1 - 3 - 4 699 81
BC055950.1 - 3 - 4 401 81
BC071266.1 - 3 - 4 603 81

View Full Product Details