Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fads3
Entrez Gene ID
60527
Gene Name
fatty acid desaturase 3
Gene Aliases
AI464531
Chromosome Location
Chr.19: 10040855 - 10059671 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021890.3 NP_068690.3 5 - 6 937 80
GenBank mRNA AB041560.1 - 5 - 6 879 80
AK029318.1 - 5 - 6 796 80
AK080414.1 - 5 - 6 789 80
AK154534.1 - 5 - 6 814 80
AK172509.1 - 5 - 6 916 80
BC055950.1 - 5 - 6 618 80
BC071266.1 - 4 - 5 925 80

View Full Product Details