Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr88
Entrez Gene ID
64378
Gene Name
G-protein coupled receptor 88
Gene Aliases
AW061286, Strg
Chromosome Location
Chr.3: 116249654 - 116253498 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022427.2 NP_071872.1 1 - 2 373 53
GenBank mRNA AB042408.1 - 1 - 2 343 53
BC029939.1 - 1 - 2 284 53

View Full Product Details