Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nit2
Entrez Gene ID
52633
Gene Name
nitrilase family, member 2
Gene Aliases
1190017B19Rik, D16Ertd502e
Chromosome Location
Chr.16: 57156665 - 57167332 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023175.1 NP_075664.1 8 - 9 730 83
GenBank mRNA AF284573.1 - 8 - 9 730 83
AK003604.1 - 5 - 6 425 83
AK004533.1 - 8 - 9 713 82
AK004535.1 - 8 - 9 716 83
BC020153.1 - 8 - 9 719 83
BC080749.1 - 2 - 3 804 83

View Full Product Details