Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chordc1
Entrez Gene ID
66917
Gene Name
cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1
Gene Aliases
1110001O09Rik, AA409036, Chp-1, morgana
Chromosome Location
Chr.9: 18292233 - 18314000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025844.2 NP_080120.1 3 - 4 291 100
GenBank mRNA AK003259.1 - 3 - 4 292 100
AK012747.1 - 3 - 4 276 100
AK132696.1 - - 271 100
BC018374.1 - 3 - 4 291 100

View Full Product Details