The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290697.1 | NP_001277626.1 | 1 - 2 | 489 | 100 |
NM_026023.5 | NP_080299.4 | 1 - 2 | 489 | 100 | |
GenBank mRNA | AK004388.1 | - | 1 - 2 | 483 | 100 |
AK005909.1 | - | 1 - 2 | 484 | 100 | |
AK010850.1 | - | 1 - 2 | 296 | 100 | |
AK012388.1 | - | 1 - 2 | 267 | 100 | |
AK031779.1 | - | 1 - 2 | 267 | 100 | |
BC005646.1 | - | 1 - 2 | 250 | 100 | |
BE949482.1 | - | 1 - 2 | 209 | 100 |