The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037846.3 | NP_001032935.2 | 15 - 16 | 1687 | 119 |
NM_001037847.2 | NP_001032936.2 | 15 - 16 | 1617 | 119 | |
NM_028082.2 | NP_082358.2 | 14 - 15 | 1564 | 119 | |
NR_024327.1 | 13 - 14 | 1497 | 119 | ||
GenBank mRNA | AK011231.1 | - | 13 - 14 | 1487 | 119 |
AK078121.1 | - | 15 - 16 | 1677 | 119 | |
AK149767.1 | - | 15 - 16 | 1661 | 119 | |
BC043133.1 | - | 15 - 16 | 1588 | 119 | |
BC063105.1 | - | 16 - 17 | 1830 | 119 | |
BC065171.1 | - | 15 - 16 | 1617 | 119 | |
BC090624.1 | - | 13 - 14 | 1493 | 119 |