The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029402.3 | NP_083678.2 | 2 - 3 | 316 | 62 |
GenBank mRNA | AK004875.2 | - | 2 - 3 | 316 | 62 |
AK016520.1 | - | 2 - 3 | 286 | 62 | |
AK028589.1 | - | 2 - 3 | 285 | 62 | |
AK144693.1 | - | 2 - 3 | 289 | 62 | |
AK160597.1 | - | 2 - 3 | 301 | 62 | |
AK220287.1 | - | 2 - 3 | 291 | 62 | |
BC026779.1 | - | 2 - 3 | 217 | 62 | |
BC027428.1 | - | 2 - 3 | 279 | 62 |