The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252625.1 | NP_001239554.1 | 5 - 6 | 804 | 55 |
NM_001252626.1 | NP_001239555.1 | 6 - 7 | 876 | 55 | |
NM_029880.3 | NP_084156.2 | 5 - 6 | 747 | 55 | |
GenBank mRNA | AK007522.1 | - | 6 - 7 | 760 | 55 |
AK020666.1 | - | 5 - 6 | 681 | 55 | |
AK021033.1 | - | 5 - 6 | 639 | 55 | |
AK036168.1 | - | 5 - 6 | 669 | 55 | |
AK145232.1 | - | 5 - 6 | 695 | 55 | |
AK159187.1 | - | 5 - 6 | 643 | 55 | |
AK159932.1 | - | 5 - 6 | 644 | 55 | |
AK168895.1 | - | 5 - 6 | 638 | 55 | |
BC021466.1 | - | 5 - 6 | 693 | 55 |