The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252625.1 | NP_001239554.1 | 6 - 7 | 1018 | 88 |
NM_001252626.1 | NP_001239555.1 | 7 - 8 | 1090 | 88 | |
NM_029880.3 | NP_084156.2 | 6 - 7 | 961 | 88 | |
GenBank mRNA | AK007522.1 | - | 7 - 8 | 974 | 88 |
AK021033.1 | - | 6 - 7 | 853 | 88 | |
AK036168.1 | - | 6 - 7 | 883 | 88 | |
AK159187.1 | - | 6 - 7 | 857 | 88 | |
AK159932.1 | - | 6 - 7 | 858 | 88 | |
AK168895.1 | - | 6 - 7 | 852 | 88 |