Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem50b
Entrez Gene ID
77975
Gene Name
transmembrane protein 50B
Gene Aliases
AU015466, AU019872, B230114J08Rik
Chromosome Location
Chr.16: 91574508 - 91597680 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030018.3 NP_084294.1 2 - 3 235 64
GenBank mRNA AK020977.1 - 2 - 3 240 64
AK098154.1 - 2 - 3 199 64
AK133966.1 - 2 - 3 263 64
AK150773.1 - - 223 64
AK151796.1 - 2 - 3 265 64
AK152190.1 - 2 - 3 251 64
AK152767.1 - 2 - 3 251 64
AK155082.1 - 2 - 3 288 64
AK170684.1 - 2 - 3 230 64
AK172335.1 - 2 - 3 223 64
BC004841.1 - 2 - 3 238 64

View Full Product Details