Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lysmd3
Entrez Gene ID
80289
Gene Name
LysM, putative peptidoglycan-binding, domain containing 3
Gene Aliases
1110030H10Rik, BC003322
Chromosome Location
Chr.13: 81657657 - 81672871 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030257.1 NP_084533.1 1 - 2 135 59
GenBank mRNA AK029514.1 - 1 - 2 113 59
BC003322.1 - 1 - 2 135 59

View Full Product Details