The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030726.1 | NP_109651.1 | 1 - 1 | 1762 | 71 |
GenBank mRNA | AF295365.1 | - | 1 - 1 | 1762 | 71 |
BC116696.1 | - | 1 - 1 | 1392 | 71 | |
BC117525.1 | - | 1 - 1 | 1390 | 71 |