The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159319.2 | NP_001152791.1 | 1 - 2 | 288 | 63 |
NM_001282977.1 | NP_001269906.1 | 1 - 2 | 288 | 63 | |
NM_001282978.1 | NP_001269907.1 | 1 - 2 | 288 | 63 | |
NM_031173.4 | NP_112450.1 | 1 - 2 | 288 | 63 | |
NM_145121.3 | NP_660099.2 | 1 - 2 | 288 | 63 | |
GenBank mRNA | AB685743.1 | - | 1 - 2 | 272 | 63 |
AF068898.1 | - | 1 - 2 | 64 | 63 | |
AF322905.1 | - | 1 - 2 | 292 | 63 | |
AK155193.1 | - | 1 - 2 | 289 | 63 | |
AY094172.1 | - | 1 - 2 | 86 | 63 | |
AY094173.1 | - | 1 - 2 | 86 | 63 | |
BC077713.1 | - | 1 - 2 | 284 | 63 | |
BY080419.1 | - | 1 - 2 | 289 | 63 |