The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032541.1 | NP_115930.1 | 1 - 2 | 124 | 72 |
GenBank mRNA | AF297664.1 | - | 1 - 2 | 124 | 72 |
AF503444.1 | - | 1 - 2 | 124 | 72 | |
BC021587.1 | - | 1 - 2 | 107 | 72 |