Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ftcd
Entrez Gene ID
14317
Gene Name
formiminotransferase cyclodeaminase
Gene Aliases
-
Chromosome Location
Chr.10: 76575648 - 76590340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080845.2 NP_543121.1 10 - 11 1321 63
GenBank mRNA AK165414.1 - 8 - 9 1528 63
BC010813.1 - 10 - 11 1283 63
BC024078.1 - 10 - 11 1315 63
CB949427.1 - 4 - 5 459 63

View Full Product Details