Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntnap4
Entrez Gene ID
170571
Gene Name
contactin associated protein-like 4
Gene Aliases
Caspr4, E130114F09Rik
Chromosome Location
Chr.8: 112569965 - 112882707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130457.2 NP_569724.2 2 - 3 297 76
GenBank mRNA AF333770.1 - 2 - 3 262 76
AK042607.1 - 2 - 3 286 76
AK053611.1 - 2 - 3 289 76
AK081891.1 - 2 - 3 290 76
AK142816.1 - 2 - 3 293 76
AK173255.1 - 2 - 3 272 76
BC034628.1 - 2 - 3 297 76

View Full Product Details