Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cacng6
Entrez Gene ID
54378
Gene Name
calcium channel, voltage-dependent, gamma subunit 6
Gene Aliases
2310033H20Rik, AW050150
Chromosome Location
Chr.7: 3423580 - 3435667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133183.1 NP_573446.1 1 - 2 331 96
GenBank mRNA AF361348.1 - 1 - 2 331 96
BC115456.1 - 1 - 2 331 96
BC115457.1 - 1 - 2 331 96

View Full Product Details