The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133183.1 | NP_573446.1 | 3 - 4 | 544 | 56 |
GenBank mRNA | AF361348.1 | - | 3 - 4 | 544 | 56 |
BC115456.1 | - | 3 - 4 | 544 | 56 | |
BC115457.1 | - | 3 - 4 | 544 | 56 |