Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prima1
Entrez Gene ID
170952
Gene Name
proline rich membrane anchor 1
Gene Aliases
B230212M13Rik, BB120497, PRiMA
Chromosome Location
Chr.12: 103194453 - 103242720 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133364.2 NP_579942.2 1 - 2 68 90
GenBank mRNA AK140034.1 - 1 - 2 69 90
AY043275.1 - 1 - 2 69 90
AY225515.1 - 1 - 2 69 90
BC134395.1 - 1 - 2 50 90
BC145454.1 - 1 - 2 48 90
BC171927.1 - 1 - 2 48 90
BC171928.1 - 1 - 2 48 90

View Full Product Details