Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Miip
Entrez Gene ID
28010
Gene Name
migration and invasion inhibitory protein
Gene Aliases
AA553001, AI853621, D4Wsu114e, IIP45
Chromosome Location
Chr.4: 147860778 - 147868719 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025365.2 NP_001020536.2 1 - 2 349 124
GenBank mRNA BC017525.1 - 1 - 2 92 124
BC024069.1 - 1 - 2 131 124
FJ618908.1 - 1 - 2 349 124

View Full Product Details