Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc28
Entrez Gene ID
246738
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 28
Gene Aliases
BC020175, ORF28
Chromosome Location
Chr.16: 91614257 - 91618999 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138664.2 NP_619605.2 1 - 2 236 88
GenBank mRNA AK081175.1 - 1 - 2 232 88
BB622009.1 - 1 - 2 236 88
BC020175.1 - 1 - 2 230 88

View Full Product Details