Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssx2ip
Entrez Gene ID
99167
Gene Name
synovial sarcoma, X breakpoint 2 interacting protein
Gene Aliases
AU014939, AU042321, Adip
Chromosome Location
Chr.3: 146404617 - 146440144 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138744.3 NP_620083.1 10 - 11 1504 88
GenBank mRNA AF532969.1 - 9 - 10 1299 88
AK173064.1 - 10 - 11 1491 88
BC021749.1 - 10 - 11 1459 88

View Full Product Details