Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ssx2ip
Entrez Gene ID
99167
Gene Name
synovial sarcoma, X breakpoint 2 interacting protein
Gene Aliases
AU014939, AU042321, Adip
Chromosome Location
Chr.3: 146404617 - 146440144 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253768.1 NP_001240697.1 14 - 15 2214 99
NM_001253769.1 NP_001240698.1 13 - 14 2110 99
NM_001253770.1 NP_001240699.1 13 - 14 1959 99
NM_138744.3 NP_620083.1 13 - 14 1962 99
GenBank mRNA AF532969.1 - 12 - 13 1757 99
AK031356.1 - 14 - 15 1938 99
AK043865.1 - 13 - 14 2063 99
AK049080.1 - 13 - 14 2120 99
AK077112.1 - 14 - 15 1999 99
AK173064.1 - 13 - 14 1949 99
BC021749.1 - 13 - 14 1917 99
BC031527.1 - 13 - 14 1925 99

View Full Product Details