The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144550.4 | NP_653133.3 | 11 - 12 | 1398 | 88 |
GenBank mRNA | AK048789.1 | - | 11 - 12 | 1342 | 88 |
AY099108.1 | - | 11 - 12 | 1678 | 88 | |
BC024956.1 | - | 3 - 4 | 752 | 88 | |
BC052381.1 | - | 11 - 12 | 1352 | 88 | |
BC056960.1 | - | 11 - 12 | 1352 | 88 |