The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144553.2 | NP_653136.2 | 1 - 2 | 299 | 65 |
GenBank mRNA | AB076696.1 | - | 1 - 2 | 299 | 65 |
AK036384.1 | - | 1 - 2 | 101 | 65 | |
AK048470.1 | - | 1 - 2 | 69 | 65 | |
AK085882.1 | - | 1 - 2 | 80 | 65 | |
AK129032.1 | - | 1 - 2 | 45 | 65 | |
AK135820.1 | - | 1 - 2 | 106 | 65 | |
AK147955.1 | - | 1 - 2 | 69 | 65 | |
AK164575.1 | - | 1 - 2 | 94 | 65 | |
BC027273.1 | - | 1 - 2 | 39 | 65 |