Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uhrf2
Entrez Gene ID
109113
Gene Name
ubiquitin-like, containing PHD and RING finger domains 2
Gene Aliases
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf
Chromosome Location
Chr.19: 30030433 - 30093729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144873.2 NP_659122.2 2 - 3 659 70
GenBank mRNA AB116653.1 - 2 - 3 394 70
AF274047.1 - 2 - 3 569 70
AK031036.1 - 2 - 3 721 70
AK041564.1 - 2 - 3 681 70
AK042321.1 - 2 - 3 700 70
AK051743.1 - 2 - 3 700 70
AK080925.1 - 2 - 3 700 70
AK172682.1 - 2 - 3 543 70
AK206406.1 - 1 - 2 105 70
BC060241.1 - 2 - 3 659 70

View Full Product Details