Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pomt1
Entrez Gene ID
99011
Gene Name
protein-O-mannosyltransferase 1
Gene Aliases
AI505244
Chromosome Location
Chr.2: 32236595 - 32255005 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145145.1 NP_660127.1 9 - 10 945 67
GenBank mRNA AK053889.1 - 8 - 9 1465 67
AK153984.1 - 9 - 10 1044 67
AY007238.1 - 9 - 10 985 67
AY494857.1 - 9 - 10 1005 67
BC027325.1 - 9 - 10 945 67

View Full Product Details