Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pqlc2
Entrez Gene ID
212555
Gene Name
PQ loop repeat containing 2
Gene Aliases
BC019216
Chromosome Location
Chr.4: 139294030 - 139310711 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145384.2 NP_663359.2 4 - 5 572 76
GenBank mRNA AK081661.1 - 4 - 5 559 76
AK156668.1 - 4 - 5 572 76
BC019216.1 - 4 - 5 543 76
BC025609.1 - 3 - 4 510 76

View Full Product Details