The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130194.1 | NP_001123666.1 | 6 - 7 | 800 | 67 |
NR_052011.1 | 6 - 7 | 1041 | 67 | ||
GenBank mRNA | AY450428.1 | - | 6 - 7 | 1042 | 67 |
BC019528.1 | - | 5 - 6 | 774 | 67 | |
BC031186.1 | - | 5 - 6 | 761 | 67 | |
BC036157.1 | - | 6 - 7 | 800 | 67 | |
BC036163.1 | - | 6 - 7 | 841 | 67 |